Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 mg | Please contact Customer Service for Availability | 11 200,00 CZK |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
ticlopidine
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
[s]1c2c(cc1)C(=O)N(CC2)Cc3c(cccc3)Cl
InChI
1S/C14H12ClNOS/c15-12-4-2-1-3-10(12)9-16-7-5-13-11(14(16)17)6-8-18-13/h1-4,6,8H,5,7,9H2
InChI key
KQQBLQBNIVSMLU-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard | description certified reference material, pharmaceutical secondary standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename EDQM | manufacturer/tradename - |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical secondary standard, certified reference material |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical |
| format neat | format neat | format neat | format - |
| API family ticlopidine | API family ticlopidine | API family - | API family - |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-30°C |
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


