Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 mg | Please contact Customer Service for Availability | 39 800,00 CZK |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
valrubicin
manufacturer/tradename
USP
application(s)
pharmaceutical
pharmaceutical (small molecule)
format
mixture
storage temp.
−20°C
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description - | description United States Pharmacopeia (USP) Reference Standard |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade - | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP |
| application(s) pharmaceutical | application(s) pharmaceutical (small molecule) | application(s) - | application(s) pharmaceutical |
| format mixture | format neat | format - | format mixture |
| storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C |
| API family valrubicin | API family valrubicin | API family - | API family etoposide |
Signal Word
Danger
Hazard Codes
Hazard Classifications
Acute Tox. 4 Oral - Carc. 1B - Muta. 1B - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Precautionary Statements
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



