Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical analytical impurity (PAI)
agency
USP
form
solid
shelf life
at -20 °C
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-30°C
General description
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description Pharmaceutical Analytical Impurity (PAI) | description ≥96.5% (HPLC) | description VETRANAL®, analytical standard |
| grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade - | grade analytical standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - |
| application(s) pharmaceutical small molecule | application(s) pharmaceutical | application(s) - | application(s) forensics and toxicology |
| shelf life at -20 °C | shelf life - | shelf life - | shelf life limited shelf life, expiry date on the label |
| format neat | format neat | format - | format neat |
| form solid | form - | form powder | form - |
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 1B - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




