Skip to Content
Merck

Skip To

1A04560

USP

Sugammadex Sodium Sulfoxide

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

Sugammadex Sodium Sulfoxide, (6A,6B,6C,6D,6E,6F,6G,6H-octadeoxy-6A,6B,6C,6D,6E,6F,6G,6H-octakis[(2-carboxyethyl)sulfinyl]-gamma-Cyclodextrin, sodium salt)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Please contact Customer Service for Availability
20 100,00 CZK

About This Item

Empirical Formula (Hill Notation):
C72H104O56S8·8Na
CAS Number:
Molecular Weight:
2306.00
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904

20 100,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical analytical impurity (PAI)

agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

-10 to -25°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.
Sugammadex Sodium Sulfoxide is a USP Pharmaceutical Analytical Impurity (PAI).


USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.


Associated Drug Substance: Sugammadex


Therapeutic Area: Neuromuscular

Application

Sugammadex Sodium Sulfoxide (USP PAI) is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:
1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1A04310SML32241A04300
description

Pharmaceutical Analytical Impurity (PAI)

description

Pharmaceutical Analytical Impurity (PAI)

description

≥95% (HPLC)

description

Pharmaceutical Analytical Impurity (PAI)

grade

pharmaceutical analytical impurity (PAI)

grade

pharmaceutical analytical impurity (PAI)

grade

-

grade

pharmaceutical analytical impurity (PAI)

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

format

neat

format

neat

format

-

format

neat

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical small molecule

application(s)

-

application(s)

pharmaceutical small molecule

agency

USP

agency

USP

agency

-

agency

USP

storage temp.

-10 to -25°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library