Skip to Content
Merck

Skip To

1A06150

USP

Atazanavir S,R,S,S-diastereomer

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

Atazanavir S,R,S,S-diastereomer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C38H52N6O7
Molecular Weight:
704.86
UNSPSC Code:
41116107
eCl@ss:
25140904
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical analytical impurity (PAI)

agency

USP

form

solid

packaging

pkg of 25 mg

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

O=C(OC)NC(C(=O)NN(CC=1C=CC(=CC1)C2=NC=CC=C2)CC(O)C(NC(=O)C(NC(=O)OC)C(C)(C)C)CC=3C=CC=CC3)C(C)(C)C

InChI

InChI=1S/C38H52N6O7/c1-37(2,3)31(41-35(48)50-7)33(46)40-29(22-25-14-10-9-11-15-25)30(45)24-44(43-34(47)32(38(4,5)6)42-36(49)51-8)23-26-17-19-27(20-18-26)28-16-12-13-21-39-28/h9-21,29-32,45H,22-24H2,1-8H3,(H,40,46)(H,41,48)(H,42,49)(H,43,47)/t29-,30+,31+,32+/m0/s1

InChI key

AXRYRYVKAWYZBR-SYEZAVJTSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Find answers to frequently asked questions on USP products in USP FAQs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
10443451A061701A06190
description

Pharmaceutical Analytical Impurity (PAI)

description

United States Pharmacopeia (USP) Reference Standard

description

Pharmaceutical Analytical Impurity (PAI)

description

Pharmaceutical Analytical Impurity (PAI)

grade

pharmaceutical analytical impurity (PAI)

grade

pharmaceutical primary standard

grade

pharmaceutical analytical impurity (PAI)

grade

pharmaceutical analytical impurity (PAI)

format

neat

format

mixture

format

neat

format

neat

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

packaging

pkg of 25 mg

packaging

-

packaging

-

packaging

pkg of 25 mg

agency

USP

agency

USP

agency

USP

agency

USP


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library