Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical analytical impurity (PAI)
agency
USP
form
powder
packaging
pkg of 25 mg
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
O=C(OCC(Cl)Cl)OC1C(=O)C2(C)C(O)CC3OCC3(OC(=O)C)C2C(OC(=O)C=4C=CC=CC4)C5(O)CC(OC(=O)C(O)C(NC(=O)OC(C)(C)C)C=6C=CC=CC6)C(=C1C5(C)C)C
InChI
InChI=1S/C46H55Cl2NO16/c1-23-27(61-39(55)33(52)32(25-15-11-9-12-16-25)49-40(56)65-42(3,4)5)20-46(58)37(63-38(54)26-17-13-10-14-18-26)35-44(8,28(51)19-29-45(35,22-60-29)64-24(2)50)36(53)34(31(23)43(46,6)7)62-41(57)59-21-30(47)48/h9-18,27-30,32-35,37,51-52,58H,19-22H2,1-8H3,(H,49,56)/t27-,28-,29+,32-,33+,34+,35-,37-,44+,45-,46+/m0/s1
InChI key
NOGDKPUOAHYBCO-OMYDMPTMSA-N
General description
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| description Pharmaceutical Analytical Impurity (PAI) | description Pharmaceutical Analytical Impurity (PAI) | description Pharmaceutical Analytical Impurity (PAI) | description European Pharmacopoeia (EP) Reference Standard |
| grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical primary standard |
| format neat | format neat | format neat | format neat |
| form powder | form powder | form powder | form - |
| packaging pkg of 25 mg | packaging pkg of 25 mg | packaging - | packaging - |
| application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) - |
| storage temp. 2-8°C | storage temp. -10 to -25°C | storage temp. 2-8°C | storage temp. 2-8°C |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Irrit. 2
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

