Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 mg | Please contact Customer Service for Availability | 52 600,00 CZK |
About This Item
52 600,00 CZK
grade
pharmaceutical analytical impurity (PAI)
agency
USP
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-30°C
General description
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| description pharmaceutical analytical impurity (PAI) | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description British Pharmacopoeia (BP) Reference Standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename BP |
| grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| application(s) pharmaceutical small molecule | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) pharmaceutical |
| format neat | format neat | format neat | format neat |
| agency USP | agency USP | agency USP | agency - |
| storage temp. 2-30°C | storage temp. 15-25°C | storage temp. 2-8°C | storage temp. 2-8°C |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Irrit. 2
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


