Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C34H34F5N3O5ClH
CAS Number:
Molecular Weight:
696.10
UNSPSC Code:
41116107
eCl@ss:
25140904
Pricing and availability is not currently available.
grade
pharmaceutical analytical impurity (PAI)
agency
USP
form
solid
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
1 of 1
This Item | ||
|---|---|---|
| description Pharmaceutical Analytical Impurity (PAI) | description Pharmaceutical Analytical Impurity (PAI) | description ≥98% (HPLC) |
| grade pharmaceutical analytical impurity (PAI) | grade pharmaceutical analytical impurity (PAI) | grade - |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename - |
| application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) - |
| form solid | form solid | form powder |
| format neat | format neat | format - |
| agency USP | agency USP | agency - |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Repr. 2
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


