Skip to Content
Merck

Skip To

1A14740

USP

Crisaborole Dimer

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

Crisaborole Dimer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C28H20N2O4
Molecular Weight:
448.47
UNSPSC Code:
41116107
eCl@ss:
25140904
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical analytical impurity (PAI)

agency

USP

form

powder

shelf life

per USP

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

format

neat

storage temp.

−20°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for research use only and not for use in diagnostic procedures.

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PZ0037FLUH9C59ED43
description

Pharmaceutical Analytical Impurity (PAI)

description

≥98% (HPLC)

description

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical analytical impurity (PAI)

grade

-

grade

-

shelf life

per USP

shelf life

-

shelf life

-

application(s)

pharmaceutical small molecule

application(s)

-

application(s)

-

format

neat

format

-

format

-

form

powder

form

powder

form

-


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library