Skip to Content
Merck

Skip To

V001063

Tropaeolin OO

Synonym(s):

4-[(4-Anilinophenyl)azo]benzenesulfonic acid sodium salt, Orange IV

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H14N3NaO3S
CAS Number:
Molecular Weight:
375.38
PubChem Substance ID:
EC Number:
209-071-4
Beilstein/REAXYS Number:
3831566
Colour Index Number:
13080
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


SMILES string

[Na+].[O-]S(=O)(=O)c1ccc(cc1)\N=N\c2ccc(Nc3ccccc3)cc2

InChI

1S/C18H15N3O3S.Na/c22-25(23,24)18-12-10-17(11-13-18)21-20-16-8-6-15(7-9-16)19-14-4-2-1-3-5-14;/h1-13,19H,(H,22,23,24);/q;+1/p-1/b21-20+;

InChI key

MLVYOYVMOZFHIU-ANVLNOONSA-M



Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library