Skip to Content
Merck

Skip To

V900147

Hexadecane

98%, solid or liquid, suitable for analytical testing, Vetec™, reagent grade

Synonym(s):

n-Hexadecane, Cetane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)14CH3
CAS Number:
Molecular Weight:
226.44
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
208-878-9
Beilstein/REAXYS Number:
1736592
MDL number:
Assay:
98%
Grade:
reagent grade
Bp:
287 °C (lit.)
Vapor pressure:
1 mmHg ( 105.3 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Hexadecane, Vetec™, reagent grade, 98%

grade

reagent grade

vapor density

7.8 (vs air)

vapor pressure

1 mmHg ( 105.3 °C)

product line

Vetec™

assay

98%

form

solid or liquid

autoignition temp.

395 °F

dilution

(for analytical testing)

refractive index

n20/D 1.434 (lit.)

bp

287 °C (lit.)

mp

18 °C (lit.)

transition temp

solidification point 17.5-18.5 °C

density

0.773 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCC

InChI

1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3

InChI key

DCAYPVUWAIABOU-UHFFFAOYSA-N

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Hexadecane


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

supp_hazards

Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

275.0 °F - closed cup

Flash Point (°C)

135 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters