Skip to Content
Merck

Skip To

V900552

γ-Valerolactone

Vetec™, reagent grade, 98%

Synonym(s):

γ-Methyl-γ-butyrolactone, 4,5-Dihydro-5-methyl-2(3H)-furanone, 4-Hydroxypentanoic acid lactone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
EC Number:
203-569-5
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
80420
MDL number:
eCl@ss:
32150319
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

reagent grade

vapor density

3.45 (vs air)

product line

Vetec™

assay

98%

refractive index

n20/D 1.432 (lit.)

bp

207-208 °C (lit.), 82-85 °C/10 mmHg (lit.)

mp

−31 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

SMILES string

CC1CCC(=O)O1

InChI

1S/C5H8O2/c1-4-2-3-5(6)7-4/h4H,2-3H2,1H3

InChI key

GAEKPEKOJKCEMS-UHFFFAOYSA-N

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under γ-Valerolactone


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

204.8 °F - closed cup

Flash Point (°C)

96 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

108-29-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters