Product Name
Atto 495 BioReagent, suitable for fluorescence, ≥98.0% (HPCE)
Product Number
16951
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Red to Very Dark Red and Red-Brown and Brown-Red
APPEARANCE (FORM)Powder
PURITY (HPCE AREA %)≥ 98.0 %
SOLUBILITY (COLOR)FOR INFORMATION
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 mg/ml in DMSO
EMISSION MAXIMUM526 ± 5 nm
REMARKS ON LUMINESCENCEEx. 499 nm in 0.1 M phosphate pH 7.0