Product Name
Eriodictyol ≥95.0% (HPLC)
Product Number
94258
Product Brand
sigma
CAS Number
4049-38-1
Molecular Weight
288.25
Test
Specification
APPEARANCE (COLOR)White to Light Beige to Light Brown
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 95.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE