Product Name
Bromoperoxidase from Corallina officinalis lyophilized powder, ≥100 units/mg protein (Lowry)
Product Number
b2170
Product Brand
sigma
CAS Number
69279-19-2
Molecular Weight
Test
Specification
APPEARANCEREPORT RESULT
PROTEIN BY LOWRYAPPROX. 10%
ENZYMATIC ACTIVITYMINIMUM 100 UNITS/MG PROTEIN
UNIT DEFINITIONONE UNIT WILL CATALYZE THE CONVERSION OF 1.0MICROMOLE OF MONCHLORODIMEDON TO MONOBROMOMONOCHLORODIMEDON PER MINUTE AT PH6.4 AT 25DEGC.
ENZYMATIC IMPURITIESCHLOROPEROXIDASE: <1%
DOCUMENT # B2170/09/06/00/1