Product Name
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
Product Number
d0431
Product Brand
sigma
CAS Number
14933-08-5
Molecular Weight
335.55
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
PURITY (TLC AREA %)≥ 99.0 %
ASSAY (ELEMENTAL ANALYSIS)≥ 97.0 %
INSOLUBLE MATTER≤ 0.1 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear to Slightly Hazy
SOLUBILITY (METHOD)1 M in Water at 20 DEG C
RESIDUE ON IGNITION≤ 0.1 %
CARBON CONTENT60.85 % (THEORY)
HYDROGEN CONTENT11.11 % (THEORY)
NITROGEN CONTENT4.17 % (THEORY)
INFRARED SPECTRUMCONFORMS TO STRUCTURE
METAL TRACE ANALYSIS (ICP)CORRESPONDS TO REQUIREMENTS
ALUMINIUM (ICP)≤ 0.0005 %
CALCIUM (ICP)≤ 0.002 %
COPPER (ICP)≤ 0.0005 %
IRON (ICP)≤ 0.0005 %
POTASSIUM (ICP)≤ 0.005 %
MAGNESIUM (ICP)≤ 0.0005 %
SODIUM (ICP)≤ 0.005 %
LEAD (ICP)≤ 0.001 %
SULFATE (ICP)≤ 0.05 %
ZINC (ICP)≤ 0.0005 %
TOTAL PHOSPHORUS AS PO4 (ICP)≤ 0.0005 %
CHLORIDE (CL)≤ 0.05 %
ION CHROMATOGRAPHY (NH4+)≤ 500 mg/kg