Product Name
2,2′-Dithiodipyridine powder
Product Number
d5767
Product Brand
sigma
CAS Number
2127-03-9
Molecular Weight
220.31
Test
Specification
APPEARANCE (COLOR)White to Light Yellow
APPEARANCE (FORM)Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
PURITY (GC AREA %)≥ 98 %
SOLUBILITY (COLOR)Colorless to Light Yellow
SOLUBILITY (TURBIDITY)Clear to Very Slightly Hazy
SOLUBILITY (METHOD)0.5G IN 10 ML MEOH
CARBON CONTENT54.52 % (Theory)
NITROGEN CONTENT12.72 % (Theory)