Product Name
1,1-Dimethyl-4-phenylpiperazinium iodide ≥98% (TLC or titration)
Product Number
d5891
Product Brand
sigma
CAS Number
54-77-3
Molecular Weight
318.20
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
ARGENTOMETR. TITRATION97.5 - 102.5 %
SOLUBILITY (COLOR)Colorless to Very Dark Yellow
SOLUBILITY (TURBIDITY)Clear to Slightly Hazy
SOLUBILITY (METHOD)50 MG/ML IN ACETIC ACID:WATER (1:1)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE