Product Name
1-Hexacosanol ≥97% (capillary GC)
Product Number
h2139
Product Brand
sigma
CAS Number
506-52-5
Molecular Weight
382.71
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
PURITY (GC AREA %)≥ 96.5 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)50 MG/ML IN ETHANOL
CARBON CONTENT78.8 - 84.4 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE