Merck
  • Xanthine Oxidoreductase Promotes The Inflammatory State Of Mononuclear Phagocytes Through Effects On Chemokine Expression, Peroxisome Proliferator-activated Receptor-{gamma} Sumoylation, And HIF-1{alpha}.
Xanthine Oxidoreductase Promotes The Inflammatory State Of Mononuclear Phagocytes Through Effects On Chemokine Expression, Peroxisome Proliferator-activated Receptor-{gamma} Sumoylation, And HIF-1{alpha}.
The Journal of Thoracic and Cardiovascular Surgery (2011)
Gibbings, S., et al.