Merck
  • Translational control of ERK signaling through miRNA/4EHP-directed silencing.
Translational control of ERK signaling through miRNA/4EHP-directed silencing.
eLife (2018-02-08)
Seyed Mehdi Jafarnejad, Clément Chapat, Edna Matta-Camacho, Idit Anna Gelbart, Geoffrey G Hesketh, Meztli Arguello, Aitor Garzia, Sung-Hoon Kim, Jan Attig, Maayan Shapiro, Masahiro Morita, Arkady Khoutorsky, Tommy Alain, Christos G Gkogkas, Noam Stern-Ginossar, Thomas Tuschl, Anne-Claude Gingras, Thomas F Duchaine, Nahum Sonenberg
ABSTRACT
MicroRNAs (miRNAs) exert a broad influence over gene expression by directing effector activities that impinge on translation and stability of mRNAs. We recently discovered that the cap-binding protein 4EHP is a key component of the mammalian miRNA-Induced Silencing Complex (miRISC), which mediates gene silencing. However, little is known about the mRNA repertoire that is controlled by the 4EHP/miRNA mechanism or its biological importance. Here, using ribosome profiling, we identify a subset of mRNAs that are translationally controlled by 4EHP. We show that the Dusp6 mRNA, which encodes an ERK1/2 phosphatase, is translationally repressed by 4EHP and a specific miRNA, miR-145. This promotes ERK1/2 phosphorylation, resulting in augmented cell growth and reduced apoptosis. Our findings thus empirically define the integral role of translational repression in miRNA-induced gene silencing and reveal a critical function for this process in the control of the ERK signaling cascade in mammalian cells.
Sigma-Aldrich
LookOut® Mycoplasma PCR Detection Kit, Optimized for use with JumpStart Taq DNA Polymerase, D9307.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid