Merck
  • The E3Σ1+ (63S1)←A3Π0+(53P1) transition in CdAr revisited: The spectrum and new analysis of the E3Σ1+ Rydberg state interatomic potential.
The E3Σ1+ (63S1)←A3Π0+(53P1) transition in CdAr revisited: The spectrum and new analysis of the E3Σ1+ Rydberg state interatomic potential.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2018-02-13)
T Urbańczyk, M Krośnicki, A Kędziorski, J Koperski
ABSTRACT
Revisited study of the E3Σ1+ (63S1)←A3Π0+(53P1) transition in CdAr using both theoretical and experimental approach is presented. Systematic detection of the E3Σ1+in,υ'←A3Π0+,υ″=6 transition frequencies with higher accuracy and spectrally narrower laser extended and improved analysis and simulation of the LIF excitation spectrum. More consistent characterization of the E3Σ1+in-Rydberg state inner well using inversed perturbation approach methodology was achieved. Free←bound transitions in the E3Σ1+in←A3Π0+,υ″=6 excitation were taken into account in the analysis and simulation of the recorded spectrum. The updated spectroscopic characterization of the A3Π0+ state was also revisited.
Sigma-Aldrich
Argon, 99.999%, Messer® CANGas