Merck
  • Caveolin-1 deficiency protects pancreatic β cells against palmitate-induced dysfunction and apoptosis.
Caveolin-1 deficiency protects pancreatic β cells against palmitate-induced dysfunction and apoptosis.
Cellular signalling (2018-03-30)
Wen Zeng, Jiansong Tang, Haicheng Li, Haixia Xu, Hongyun Lu, Hangya Peng, Chuwen Lin, Rili Gao, Shuo Lin, Keyi Lin, Kunying Liu, Yan Jiang, Jianping Weng, Longyi Zeng
ABSTRACT
Lipotoxicity leads to insulin secretion deficiency, which is among the important causes for the onset of type 2 diabetes mellitus. Thus, the restoration of β-cell mass and preservation of its endocrine function are long-sought goals in diabetes research. Previous studies have suggested that the membrane protein caveolin-1 (Cav-1) is implicated in β-cell apoptosis and insulin secretion, however, the underlying mechanisms still remains unclear. Our objective is to explore whether Cav-1 depletion protects pancreatic β cells from lipotoxicity and what are the underlying mechanisms. In this study, we found that Cav-1 silencing significantly promoted β-cell proliferation, inhibited palmitate (PA)-induced pancreatic β-cell apoptosis and enhanced insulin production and secretion. These effects were associated with enhanced activities of Akt and ERK1/2, which in turn downregulated the expression of cell cycle inhibitors (FOXO1, GSK3β, P21, P27 and P53) and upregulated the expression of Cyclin D2 and Cyclin D3. Subsequent inhibition of PI3K/Akt and ERK/MAPK pathways abolished Cav-1 depletion induced β-cell mass protection. Furthermore, under PA induced endoplasmic reticulum (ER) stress, Cav-1 silencing significantly reduced eIF2α phosphorylation and the expression of ER stress-responsive markers BiP and CHOP, which are among the known sensitizers of lipotoxicity. Our findings suggest Cav-1 as potential target molecule in T2DM treatment via the preservation of lipotoxicity-induced β-cell mass reduction and the attenuation of insulin secretion dysfunction.
Product Description
Supelco
Picric acid solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
L-(−)-Glucose, ≥99%