Merck
  • Oxygen-Evolving Mesoporous Organosilica Coated Prussian Blue Nanoplatform for Highly Efficient Photodynamic Therapy of Tumors.
Oxygen-Evolving Mesoporous Organosilica Coated Prussian Blue Nanoplatform for Highly Efficient Photodynamic Therapy of Tumors.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2018-06-08)
Zhen Lu Yang, Wei Tian, Qing Wang, Ying Zhao, Yun Lei Zhang, Ying Tian, Yu Xia Tang, Shou Ju Wang, Ying Liu, Qian Qian Ni, Guang Ming Lu, Zhao Gang Teng, Long Jiang Zhang
ABSTRACT
Oxygen (O2) plays a critical role during photodynamic therapy (PDT), however, hypoxia is quite common in most solid tumors, which limits the PDT efficacy and promotes the tumor aggression. Here, a safe and multifunctional oxygen-evolving nanoplatform is costructured to overcome this problem. It is composed of a prussian blue (PB) core and chlorin e6 (Ce6) anchored periodic mesoporous organosilica (PMO) shell (denoted as PB@PMO-Ce6). In the highly integrated nanoplatform, the PB with catalase-like activity can catalyze hydrogen peroxide to generate O2, and the Ce6 transform the O2 to generate more reactive oxygen species (ROS) upon laser irradiation for PDT. This PB@PMO-Ce6 nanoplatform presents well-defined core-shell structure, uniform diameter (105 ± 12 nm), and high biocompatibility. This study confirms that the PB@PMO-Ce6 nanoplatform can generate more ROS to enhance PDT than free Ce6 in cellular level (p < 0.001). In vivo, the singlet oxygen sensor green staining, tumor volume of tumor-bearing mice, and histopathological analysis demonstrate that this oxygen-evolving nanoplatform can elevate singlet oxygen to effectively inhibit tumor growth without obvious damage to major organs. The preliminary results from this study indicate the potential of biocompatible PB@PMO-Ce6 nanoplatform to elevate O2 and ROS for improving PDT efficacy.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)