Merck
  • Chronic Adolescent CDPPB Treatment Alters Short-Term, but not Long-Term, Glutamatergic Receptor Expression.
Chronic Adolescent CDPPB Treatment Alters Short-Term, but not Long-Term, Glutamatergic Receptor Expression.
Neurochemical research (2018-06-25)
Jeremy S Lum, Samuel J Millard, Elisabeth Frank, Natalie Matosin, Xu-Feng Huang, Lezanne Ooi, Kelly A Newell
ABSTRACT
Dysfunction of the glutamatergic system is believed to underlie many neurodevelopmental disorders including autism, Rett syndrome and schizophrenia. Metabotropic glutamate receptor (mGluR5) positive allosteric modulators (PAM) potentiate glutamatergic signaling, particularly indirectly via the NMDA receptor. Preclinical studies report mGluR5 PAMs can improve schizophrenia-relevant behaviours. Furthermore, adolescent administration has shown to prevent cognitive induced deficits in adult rodents. However, there is limited understanding of the short- and long-term neurochemical effects of mGluR5 PAMs, which may underlie their therapeutic effects. We examined the effect of 7-day adolescent (PN28-34) treatment with the mGluR5 PAM, CDDPB (30 mg/kg), on glutamatergic receptor expression at adolescence (PN35) and adulthood (PN96). Immunoblot analysis revealed that 7-day adolescent CDPPB treatment increased protein expression of glutamatergic receptors including the NMDA receptor subunits, NR1 and NR2A and the AMPA subunits (GluA1 and GluA2) in the adolescent hippocampus, changes that did not extend to adulthood. In contrast, there were no changes in the adolescent frontal cortex, however elevated mGluR5 protein expression was observed at adulthood following adolescent CDPPB treatment. The present study indicates adolescent CDPPB treatment may cause brain region dependent effects on the glutamatergic system, which do not persist into adulthood. These findings may have implications for the preclinical development of mGluR5 PAMs for the treatment of neurodevelopmental disorders.
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 7E3-1B8, clone 7E3-1B8, Chemicon®, from mouse
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-NMDAR2D Antibody, ascites fluid, clone 1G9.39A5, Chemicon®
Sigma-Aldrich
Anti-NMDAR1 Antibody, clone 54.1, clone 54.1, Chemicon®, from mouse