Merck
  • Rapid Colorimetric Detection of Cartap Residues by AgNP Sensor with Magnetic Molecularly Imprinted Microspheres as Recognition Elements.
Rapid Colorimetric Detection of Cartap Residues by AgNP Sensor with Magnetic Molecularly Imprinted Microspheres as Recognition Elements.
Molecules (Basel, Switzerland) (2018-06-15)
Mao Wu, Huiyun Deng, Yajun Fan, Yunchu Hu, Yaping Guo, Lianwu Xie
ABSTRACT
The overuse of cartap in tea tree leads to hazardous residues threatening human health. A colorimetric determination was established to detect cartap residues in tea beverages by silver nanoparticles (AgNP) sensor with magnetic molecularly imprinted polymeric microspheres (Fe₃O₄@mSiO₂@MIPs) as recognition elements. Using Fe₃O₄ as supporting core, mesoporous SiO₂ as intermediate shell, methylacrylic acid as functional monomer, and cartap as template, Fe₃O₄@mSiO₂@MIPs were prepared to selectively and magnetically separate cartap from tea solution before colorimetric determination by AgNP sensors. The core-shell Fe₃O₄@mSiO₂@MIPs were also characterized by FT-IR, TEM, VSM, and experimental adsorption. The Fe₃O₄@mSiO₂@MIPs could be rapidly separated by an external magnet in 10 s with good reusability (maintained 95.2% through 10 cycles). The adsorption process of cartap on Fe₃O₄@mSiO₂@MIPs conformed to Langmuir adsorption isotherm with maximum adsorption capacity at 0.257 mmol/g and short equilibrium time of 30 min at 298 K. The AgNP colorimetric method semi-quantified cartap ≥5 mg/L by naked eye and quantified cartap 0.1⁻5 mg/L with LOD 0.01 mg/L by UV-vis spectroscopy. The AgNP colorimetric detection after pretreatment with Fe₃O₄@mSiO₂@MIPs could be successfully utilized to recognize and detect cartap residues in tea beverages.