Merck
  • Synthesis and biological evaluation of (R)-N-(diarylmethylthio/sulfinyl)ethyl/propyl-piperidine-3-carboxylic acid hydrochlorides as novel GABA uptake inhibitors.
Synthesis and biological evaluation of (R)-N-(diarylmethylthio/sulfinyl)ethyl/propyl-piperidine-3-carboxylic acid hydrochlorides as novel GABA uptake inhibitors.
Bioorganic & medicinal chemistry letters (2007-05-23)
Jiange Zhang, Pei Zhang, Xianbo Liu, Kai Fang, Guoqiang Lin
ABSTRACT
A series of new (R)-1-(2-diarylmethylthio/sulfinyl)ethyl-piperidine-3-carboxylic acid hydrochlorides 5a-d/6a-d and (R)-1-(3-diarylmethylthio)propyl-piperidine-3-carboxylic acid hydrochlorides 5'a-d were synthesized and evaluated as gamma-aminobutyric acid uptake inhibitors through cultured cell lines expressing mouse GAT1. Biological screening results demonstrated that the compounds 6a-d with diarylmethylsulfinyl ethyl side chain show more potent GAT1 inhibitory activities than 5a-d/5'a-d with diarylmethylthio ethyl/propyl moieties. Some of them, such as 6a, exhibited excellent inhibitions of [(3)H]-GABA uptake in cultured cells, which is 496-fold higher than (R)-nipecotic acid and 11.5 times less than tiagabine. The synthesis and structure-activity relationships are discussed.
Sigma-Aldrich
(S)-(+)-3-Piperidinecarboxylic acid, 97%
Sigma-Aldrich
(R)-(–)-3-Piperidinecarboxylic acid, 97%