Merck
  • Isopeptide and ester bond ubiquitination both regulate degradation of the human dopamine receptor 4.
Isopeptide and ester bond ubiquitination both regulate degradation of the human dopamine receptor 4.
The Journal of biological chemistry (2017-11-05)
Jennifer C Peeler, Sophia Schedin-Weiss, Mariluz Soula, Manija A Kazmi, Thomas P Sakmar
ABSTRACT
How an optimal level of human dopamine D4 receptor (hD4R) is maintained in synaptic membranes is not known. We show here that hD4R is ubiquitinated in primary neurons. We go on to show that ubiquitin is attached to hD4R through isopeptide and ester bonds. When lysine (Lys) residues of the hD4R are substituted with arginine (Arg) residues, cellular hD4R protein levels increase. A synergistic effect on hD4R levels is noted when cytoplasmic serine (Ser) and threonine (Thr) residues are mutated. Chloroquine, an inhibitor of lysosomal degradation, did not have an effect on hD4R protein levels. However, treatment with bortezomib, an inhibitor of the 20S proteasome, caused a dose-dependent increase in hD4R protein levels. The effect of bortezomib was attenuated in the receptor variants that lacked Lys or Ser/Thr residues, and the hD4R mutant that lacked 17 cytoplasmic Lys, Ser, and Thr residues was nearly insensitive to bortezomib treatment. We conclude that both isopeptide and ester bond ubiquitination regulate proteasomal degradation of hD4R.
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit