Merck
  • Proteomic screening identifies the zonula occludens protein ZO-1 as a new partner for ADAM12 in invadopodia-like structures.
Proteomic screening identifies the zonula occludens protein ZO-1 as a new partner for ADAM12 in invadopodia-like structures.
Oncotarget (2018-05-17)
Bassil Dekky, Michael Ruff, Dominique Bonnier, Vincent Legagneux, Nathalie Théret
ABSTRACT
The epithelial mesenchymal transition (EMT) is a key process for cancer cell invasion and migration. This complex program whereby epithelial tumor cells loose polarity and acquire mesenchymal phenotype is driven by the regulation of cell-cell adhesion and cell-substrate interactions. We recently described the association of ADAM12 with EMT and we now use immunoprecipitation and proteomic approaches to identify interacting partners for ADAM12 during EMT. We identify twenty proteins that are involved in molecular mechanisms associated with adhesion/invasion processes. Integrative network analyses point out the zonula occludens protein ZO-1, as a new potential partner for ADAM12. In silico screening demonstrates that ZO-1 and ADAM12 are coexpressed in breast cancer cell lines sharing EMT signature. We validate the interaction between ZO-1 and ADAM12 in invasive breast cancer cell lines and show that ZO-1 and ADAM12 co-localize in actin- and cortactin-rich structures. Silencing either ADAM12 or ZO-1 inhibits gelatin degradation demonstrating that both proteins are required for matrix degradation. We further show that matrix metalloprotease 14, known to mediate degradation of collagen in invadopodia-like structures interacts with ZO-1. Depletion of PKCε that regulates the recruitment of ADAM12 and ZO-1 to cell membranes induces a decrease in ADAM12 and ZO-1 at invadopodia-like structures and degradation activity. Together our data provide evidence for a new interaction between ADAM12, a mesenchymal marker induced during TGF-β-dependent EMT and ZO-1, a scaffolding protein expressed in tight junctions of epithelial cells, both proteins being redistributed at the invadopodia-like structures of mesenchymal invasive cells to promote PKCε-dependent matrix degradation.
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit