Merck
  • Kartogenin inhibits pain behavior, chondrocyte inflammation, and attenuates osteoarthritis progression in mice through induction of IL-10.
Kartogenin inhibits pain behavior, chondrocyte inflammation, and attenuates osteoarthritis progression in mice through induction of IL-10.
Scientific reports (2018-09-16)
Ji Ye Kwon, Seung Hoon Lee, Hyun-Sik Na, KyungAh Jung, JeongWon Choi, Keun Hyung Cho, Chang-Yong Lee, Seok Jung Kim, Sung-Hwan Park, Dong-Yun Shin, Mi-La Cho
ABSTRACT
Osteoarthritis (OA) is a major degenerative joint condition that causes articular cartilage destruction. It was recently found that enhancement of chondroclasts and suppression in Treg cell differentiation are involved in the pathogenesis of OA. Kartogenin (KGN) is a small drug-like molecule that induces chondrogenesis in mesenchymal stem cells (MSCs). This study aimed to identify whether KGN can enhance severe pain behavior and improve cartilage repair in OA rat model. Induction of OA model was loaded by IA-injection of MIA. In the OA rat model, treatment an intra-articular injection of KGN. Pain levels were evaluated by analyzing PWL and PWT response in animals. Histological analysis and micro-CT images of femurs were used to analyze cartilage destruction. Gene expression was measured by real-time PCR. Immunohistochemistry was analyzed to detect protein expression. KGN injection significantly decreased pain severity and joint destruction in the MIA-induced OA model. KGN also increased mRNA levels of the anti-inflammatory cytokine IL-10 in OA patients' chondrocytes stimulated by IL-1β. Decreased chondroclast expression, and increased Treg cell expression. KGN revealed therapeutic activity with the potential to reduce pain and improve cartilage destruction. Thus, KGN could be a therapeutic molecule for OA that inhibits cartilage damage.
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)