Merck
  • A stable meta-carborane enables the generation of boron-rich peptide agonists targeting the ghrelin receptor.
A stable meta-carborane enables the generation of boron-rich peptide agonists targeting the ghrelin receptor.
Journal of peptide science : an official publication of the European Peptide Society (2018-09-01)
Dennis J Worm, Sylvia Els-Heindl, Martin Kellert, Robert Kuhnert, Stefan Saretz, Johannes Koebberling, Bernd Riedl, Evamarie Hey-Hawkins, Annette G Beck-Sickinger
ABSTRACT
Boron neutron capture therapy (BNCT) is a binary cancer therapy, which combines the biochemical targeting of a boron-containing drug with the regional localization of radiation treatment. Although the concept of BNCT has been known for decades, the selective delivery of boron into tumor cells remains challenging. G protein-coupled receptors that are overexpressed on cancer cells in combination with peptidic ligands can be potentially used as shuttle system for a tumor-directed boron uptake. In this study, we present the generation of short, boron-rich peptide conjugates that target the ghrelin receptor. Expression of the ghrelin receptor on various cancer cells makes it a viable target for BNCT. We designed a novel hexapeptide super-agonist that was modified with different specifically synthesized carborane monoclusters and tested for ghrelin receptor activation. A meta-carborane building block with a mercaptoacetic acid linker was found to be optimal for peptide modification, owing to its chemical stability and a suitable activation efficacy of the conjugate. The versatility of this carborane for the development of peptidic boron delivery agents was further demonstrated by the generation of highly potent, boron-loaded conjugates using the backbone of the known ghrelin receptor ligands growth hormone releasing peptide 6 and Ipamorelin.
Sigma-Aldrich
O-(Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate, ≥97.0% (N)
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder