Merck
  • Janus Liposomes: Gel-Assisted Formation and Bioaffinity-Directed Clustering.
Janus Liposomes: Gel-Assisted Formation and Bioaffinity-Directed Clustering.
Langmuir : the ACS journal of surfaces and colloids (2018-06-01)
Mingming Wang, Zening Liu, Wei Zhan
ABSTRACT
This article reports a high-yield procedure for preparing microsized (giant) Janus liposomes via gel-assisted lipid swelling and clustering behavior of these liposomes directed by biotin-avidin affinity binding. Confocal fluorescence microscopy reveals in detail that these new lipid colloidal particles display broken symmetry and heterogeneous surface chemistry similar to other types of Janus particles. An optimized formation procedure is presented, which reproducibly yields large liposome populations dominated by a single-domain configuration. This work further demonstrates that biotin-conjugated 1,2-dioleoyl- sn-glycero-3-phosphoethanolamine preferentially partitions into the liquid-disordered phase of the lipid matrix, rendering these Janus liposomes asymmetrical binding capacity toward avidin. This affinity binding drives irreversible and domain-specific cluster formation among Janus liposomes, whose structure and size are found to depend on the domain configuration of individual liposomes and incubation time.
Product Description
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
16:0 Biotinyl Cap PE, Avanti Research - A Croda Brand 870277P, powder
Avanti
TopFluor Cholesterol, Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
18:1 Biotinyl PE, Avanti Research - A Croda Brand 870282C
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1 Biotinyl PE, Avanti Research - A Croda Brand 870282P, powder
Avanti
18:1 Biotinyl Cap PE, Avanti Research - A Croda Brand 870273C
Avanti
18:1 Biotinyl Cap PE, Avanti Research - A Croda Brand 870273P, powder
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand