Merck
  • 3D bioprinting of BM-MSCs-loaded ECM biomimetic hydrogels for in vitro neocartilage formation.
3D bioprinting of BM-MSCs-loaded ECM biomimetic hydrogels for in vitro neocartilage formation.
Biofabrication (2016-07-20)
Marco Costantini, Joanna Idaszek, Krisztina Szöke, Jakub Jaroszewicz, Mariella Dentini, Andrea Barbetta, Jan E Brinchmann, Wojciech Święszkowski
ABSTRACT
In this work we demonstrate how to print 3D biomimetic hydrogel scaffolds for cartilage tissue engineering with high cell density (>10(7) cells ml(-1)), high cell viability (85 ÷ 90%) and high printing resolution (≈100 μm) through a two coaxial-needles system. The scaffolds were composed of modified biopolymers present in the extracellular matrix (ECM) of cartilage, namely gelatin methacrylamide (GelMA), chondroitin sulfate amino ethyl methacrylate (CS-AEMA) and hyaluronic acid methacrylate (HAMA). The polymers were used to prepare three photocurable bioinks with increasing degree of biomimicry: (i) GelMA, (ii) GelMA + CS-AEMA and (iii) GelMA + CS-AEMA + HAMA. Alginate was added to the bioinks as templating agent to form stable fibers during 3D printing. In all cases, bioink solutions were loaded with bone marrow-derived human mesenchymal stem cells (BM-MSCs). After printing, the samples were cultured in expansion (negative control) and chondrogenic media to evaluate the possible differentiating effect exerted by the biomimetic matrix or the synergistic effect of the matrix and chondrogenic supplements. After 7, 14, and 21 days, gene expression of the chondrogenic markers (COL2A1 and aggrecan), marker of osteogenesis (COL1A1) and marker of hypertrophy (COL10A1) were evaluated qualitatively by means of fluorescence immunocytochemistry and quantitatively by means of RT-qPCR. The observed enhanced viability and chondrogenic differentiation of BM-MSCs, as well as high robustness and accuracy of the employed deposition method, make the presented approach a valid candidate for advanced engineering of cartilage tissue.
Sigma-Aldrich
TissueFab® - low endotoxin GelMA-UV bioink, 0.2 μm filtered, suitable for 3D bioprinting applications
Sigma-Aldrich
TissueFab® bioink , (GelAlgHA)MA Vis/405 nm, low endotoxin
Sigma-Aldrich
TissueFab® bioink Bone UV/365 nm
Sigma-Aldrich
TissueFab® bioink Alg(Gel)ma -UV/365 nm
Sigma-Aldrich
TissueFab® bioink , (Gel)ma -UV/365 nm
Sigma-Aldrich
TissueFab® bioink , Alg(Gel)ma -Vis/525 nm
Sigma-Aldrich
TissueFab® bioink , Alg(Gel)ma -UV/365 nm
Sigma-Aldrich
TissueFab® bioink , (Gel)ma -VIS/405nm, low endotoxin
Sigma-Aldrich
TissueFab® bioink Bone, Vis/405 nm
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Fibrin (UV/365), low endotoxin
Sigma-Aldrich
TissueFab® bioink kit, Fibronectin-UV/365nm
Sigma-Aldrich
TissueFab® PODS Growth Factor Loaded Bioink Kit, Vascularization, Vis/405nm
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Fibrin (Vis/405), low endotoxin
Sigma-Aldrich
TissueFab® bioink Bone, UV/365 nm
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Laminin -UV/365 nm, low endotoxin
Sigma-Aldrich
TissueFab® bioink Bone Vis/405 nm
Sigma-Aldrich
TissueFab® GelAlg − LAP Bioink, low endotoxin, 0.2 μm filtered, suitable for 3D bioprinting applications
Sigma-Aldrich
TissueFab® bioink kit, (Gel)ma Laminin -Vis/405 nm, low endotoxin