Merck
Production of high oleic/low linoleic rice by genome editing.
Plant physiology and biochemistry : PPB (2018-05-08)
Kiyomi Abe, Etsuko Araki, Yasuhiro Suzuki, Seiichi Toki, Hiroaki Saika
ABSTRACT
Rice bran oil (RBO) contains many valuable healthy constituents, including oleic acid. Improvement of the fatty acid composition in RBO, including an increase in the content of oleic acid, which helps suppress lifestyle disease, would increase health benefits. The enzyme fatty acid desaturase 2 (FAD2) catalyzes the conversion of oleic acid to linoleic acid in plants, and FAD2 mutants exhibit altered oleic and linoleic acid content in many crops. There are three functional FAD2 genes in the genome of rice (Oryza sativa L.), and, of these, expression of the OsFAD2-1 gene is highest in rice seeds. In order to produce high oleic/low linoleic RBO, we attempted to disrupt the OsFAD2-1 gene by CRISPR/Cas9-mediated targeted mutagenesis. We succeeded in the production of homozygous OsFAD2-1 knockout rice plants. The content of oleic acid increased to more than twice that of wild type, and, surprisingly, linoleic acid, a catabolite of oleic acid by FAD2, decreased dramatically to undetectable levels in fad2-1 mutant brown rice seeds. In this study, by genome editing based on genome information, we succeeded in the production of rice whose fatty acid composition is greatly improved. We suggest that CRISPR/Cas9-mediated mutagenesis of a major gene that shows dominant expression in the target tissue could be a powerful tool to improve target traits in a tissue-specific manner.