Merck
  • α-Linolenic acid prevents hepatic steatosis and improves glucose tolerance in mice fed a high-fat diet.
α-Linolenic acid prevents hepatic steatosis and improves glucose tolerance in mice fed a high-fat diet.
Clinics (Sao Paulo, Brazil) (2018-11-01)
Natália Bonissi Gonçalves, Rafael Ferraz Bannitz, Bruna Ramos Silva, Danielle Duran Becari, Carolina Poloni, Patrícia Moreira Gomes, Milton Cesar Foss, Maria Cristina Foss-Freitas
ABSTRACT
Dietary omega-3 fatty acids have been efficacious in decreasing serum cholesterol levels and reducing the risk of cardiovascular disease. However, the metabolic and molecular changes induced by the omega-3 fatty acid α-linolenic acid (ALA), which is found in linseed oil, are not fully understood. In this study, we showed a correlation between ALA and insulin resistance, inflammation and endoplasmic reticulum stress (ERS). We studied 40 male mice (C57/BL6) divided into 4 groups: a control (C) group, a control + omega-3/ALA (CA) group, a high-fat diet (HFD) (H) group and a high-fat diet + omega-3/ALA (HA) group. For 8 weeks, the animals in the H and HA groups were fed a high-fat (60%) diet, while the animals in the C and CA groups received regular chow. The diets of the CA and HA groups were supplemented with 10% lyophilized ALA. ALA supplementation improved glucose tolerance and reduced insulin resistance, as measured by intraperitoneal glucose tolerance tests and the homeostasis model assessment for insulin resistance, respectively. In addition, ALA reduced hepatic steatosis and modified the standard fat concentration in the liver of animals fed an HFD. Dietary ALA supplementation reduced the serum levels of interleukin 6 (IL-6), interleukin 1 beta (IL-1β) and monocyte chemoattractant protein-1 (MCP-1), increased the expression of important chaperones such as binding immunoglobulin protein (BIP) and heat shock protein 70 (HSP70) and reduced the expression of C/EBP-homologous protein (CHOP) and X-box binding protein 1 (XBP1) in hepatic tissues, suggesting an ERS adaptation in response to ALA supplementation. Dietary ALA supplementation is effective in preventing hepatic steatosis; is associated with a reduction in insulin resistance, inflammation and ERS; and represents an alternative for improving liver function and obtaining metabolic benefits.
Sigma-Aldrich
Bicinchoninic Acid Kit for Protein Determination, for 200-1000 μg/ml protein
Pricing and availability is not currently available.
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Pricing and availability is not currently available.
Supelco
Bicinchoninic Acid solution
Pricing and availability is not currently available.