Merck
  • Dietary Crystalline DL-Methionine Supplementation of Oilseed Meal-Based Diets: Effects on Growth and Feed Utilization of Juvenile Nile Tilapia (Oreochromis niloticus L.)
Dietary Crystalline DL-Methionine Supplementation of Oilseed Meal-Based Diets: Effects on Growth and Feed Utilization of Juvenile Nile Tilapia (Oreochromis niloticus L.)
American Journal of Experimental Agriculture (2014)
Agbo NW, et al.