Merck
  • Epinephrine Evokes Renalase Secretion via a-Adrenoceptor/NF-kappaB Pathways in Renal Proximal Tubular Epithelial Cells
Epinephrine Evokes Renalase Secretion via a-Adrenoceptor/NF-kappaB Pathways in Renal Proximal Tubular Epithelial Cells
Kidney & Blood Pressure Research (2014)
Wang F, et al.