Merck
  • Mutations in the pancreatic secretory enzymes CPA1 and CPB1 are associated with pancreatic cancer
Mutations in the pancreatic secretory enzymes CPA1 and CPB1 are associated with pancreatic cancer
Proceedings of the National Academy of Sciences of the USA (2018)
Tamura K, et al.