Merck
  • Synthesis and antibacterial activity of 3-O-acyl-6-O-carbamoyl erythromycin A derivatives.
Synthesis and antibacterial activity of 3-O-acyl-6-O-carbamoyl erythromycin A derivatives.
Bioorganic & medicinal chemistry letters (2005-11-18)
Bin Zhu, Brett A Marinelli, Darren Abbanat, Barbara D Foleno, Todd C Henninger, Karen Bush, Mark J Macielag
ABSTRACT
A series of 3-O-acyl-6-O-carbamoyl erythromycin A derivatives has been synthesized. Several functional groups were identified as the optimal C3-substituents, and the best compounds in this series possess potent in vitro antibacterial activity against erythromycin-susceptible and erythromycin-resistant bacteria.
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg