Merck
  • Synthesis of unsymmetrical 3,4-diaryl-3-pyrrolin-2-ones utilizing pyrrole Weinreb amides.
Synthesis of unsymmetrical 3,4-diaryl-3-pyrrolin-2-ones utilizing pyrrole Weinreb amides.
The Journal of organic chemistry (2011-09-15)
Jessica G Greger, Sarah J P Yoon-Miller, Nathan R Bechtold, Scott A Flewelling, Jacob P MacDonald, Catherine R Downey, Eric A Cohen, Erin T Pelkey
ABSTRACT
A regiocontrolled synthesis of unsymmetrical 3,4-diaryl-3-pyrrolin-2-ones has been achieved in three steps from 1,2-diaryl-1-nitroethenes with pyrrole-2-carboxamides (pyrrole Weinreb amides) serving as the key linchpin intermediates. Two different methods for the preparation of the requisite nitroalkenes were investigated: (1) modified Henry reaction between arylnitromethanes and arylimines; and (2) Suzuki-Miyaura cross-coupling reaction of 2-aryl-1-bromo-1-nitroethenes with arylboronic acids. Some difficulty was encountered in the preparation of arylnitromethanes, thus leading to the exploration of a cross-coupling strategy that proved more useful. A Barton-Zard pyrrole cyclocondensation reaction between 1,2-diaryl-1-nitroethenes and N-methoxy-N-methyl-2-isocyanoacetamide gave the corresponding pyrrole Weinreb amides, which were then converted into the desired 3-pyrrolin-2-ones in two steps. Overall, this method allowed for the construction of 3,4-diaryl-3-pyrrolin-2-ones with complete regiocontrol of the substituents with respect to the lactam carbonyl. The utility of this synthetic methodology was demonstrated by the preparation of eight unsymmetrical and symmetrical 3,4-diaryl-3-pyrrolin-2-ones including the N-H lactam analogue of the selective COX-II inhibitor, rofecoxib.
Sigma-Aldrich
4-(Methanesulfonyl)phenylboronic acid, ≥95.0%