Merck
  • Templated synthesis of glycoluril hexamer and monofunctionalized cucurbit[6]uril derivatives.
Templated synthesis of glycoluril hexamer and monofunctionalized cucurbit[6]uril derivatives.
Journal of the American Chemical Society (2011-10-06)
Derick Lucas, Tsuyoshi Minami, Greg Iannuzzi, Liping Cao, James B Wittenberg, Pavel Anzenbacher, Lyle Isaacs
ABSTRACT
We report that the p-xylylenediammonium ion (11) acts as a template in the cucurbit[n]uril forming reaction that biases the reaction toward the production of methylene bridged glycoluril hexamer (6C) and bis-nor-seco-CB[10]. Hexamer 6C is readily available on the gram scale by a one step synthetic procedure that avoids chromatography. Hexamer 6C undergoes macrocylization with (substituted) phthalaldehydes 12, 14, 15, and 18-in 9 M H(2)SO(4) or concd HCl at room temperature to deliver monofunctionalized CB[6] derivatives 13, 16, 17, and 19-that are poised for further functionalization reactions. The kinetics of the macrocyclization reaction between hexamer and formaldehyde or phthalaldehyde depends on the presence and identity of ammonium ions as templates. p-Xylylenediammonium ion (11) which barely fits inside CB[6] sized cavities acts as a negative template which slows down transformation of 6C and paraformaldehyde into CB[6]. In contrast, 11 and hexanediammonium ion (20) act as a positive template that promotes the macrocyclization reaction between 6C and 12 to deliver (±)-21 as a key intermediate along the mechanistic pathway to CB[6] derivatives. Naphthalene-CB[6] derivative 19 which contains both fluorophore and ureidyl C═O metal-ion (e.g., Eu(3+)) binding sites forms the basis for a fluorescence turn-on assay for suitable ammonium ions (e.g., hexanediammonium ion and histamine).
Sigma-Aldrich
Cucurbit[6]uril hydrate, contains acid of crystalization