- beta3-Adrenoceptor agonists and (antagonists as) inverse agonists: history, perspective, constitutive activity, and stereospecific binding
beta3-Adrenoceptor agonists and (antagonists as) inverse agonists: history, perspective, constitutive activity, and stereospecific binding
Methods in Enzymology (2010)