Merck
  • Heterogeneous intrastriatal pattern of proteins regulating axon growth in normal adult human brain.
Heterogeneous intrastriatal pattern of proteins regulating axon growth in normal adult human brain.
Neurobiology of disease (2010-10-30)
Junchao Tong, Yoshiaki Furukawa, Allan Sherwin, Oleh Hornykiewicz, Stephen J Kish
ABSTRACT
There is much controversy regarding the extent of axon regeneration/sprouting ability in adult human brain. However, intrinsic differences in axon/neurite growth capability amongst striatal (caudate, putamen, nucleus accumbens) subdivisions could conceivably underlie, in part, their differential vulnerability in degenerative human brain disorders. To establish whether the distribution of axon growth markers in mature human striatum might be uniform or heterogeneous, we measured the intra-striatal pattern, in autopsied brain of normal subjects (n=40, age 18-99), of proteins involved in regulating axon growth. These proteins included polysialylated neural cell adhesion molecule (PSA-NCAM), microtubule-associated proteins TUC-4 (TOAD/Ulip/CRAMP-4) and doublecortin (DCX), and Bcl-2. The distribution of the marker proteins within the striatum was heterogeneous and inversely related to the pattern of dopamine loss previously characterized in Parkinson's disease (PD), with levels in nucleus accumbens>caudate>putamen, ventral>dorsal, and rostral putamen>caudal. In contrast, distribution of glial markers including glial fibrillary acidic protein (GFAP) and human leukocyte antigens (HLA-DRα and HLA-DR/DQ/DPβ), other Bcl-2 family proteins, and control proteins neuron-specific enolase and α-tubulin in the striatum was either homogeneous or had a pattern unmatched to dopamine loss in PD. The putamen also showed more marked age-dependent decreases in concentrations of PSA-NCAM, TUC-4, and DCX and increases in GFAP levels than caudate. We conclude that the intrastriatal pattern of several key axon growth proteins is heterogeneous in adult human brain. Further investigation will be required to establish whether this pattern, which was inversely correlated with the pattern of dopamine loss in PD, is involved to any extent in the pathophysiology of this degenerative disorder.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Glial Fibrillary Acidic Protein, porcine
Sigma-Aldrich
Anti-TUC-4 Protein Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Bak Antibody, NT, Upstate®, from rabbit
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-Bim Antibody, internal epitope, pan-Bim isoforms, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Polysialic Acid-NCAM Antibody, clone 2-2B, ascites fluid, clone 2-2B, Chemicon®