Merck
  • Time-dependent conformational changes in adsorbed albumin and its effect on platelet adhesion.
Time-dependent conformational changes in adsorbed albumin and its effect on platelet adhesion.
Langmuir : the ACS journal of surfaces and colloids (2011-12-24)
Balakrishnan Sivaraman, Robert A Latour
ABSTRACT
Recent studies have shown that platelets can adhere to adsorbed albumin (Alb) through a receptor-mediated mechanism, but only if the Alb undergoes more than a critical degree of adsorption-induced unfolding. The objectives of this research were to investigate whether Alb that was initially adsorbed in a manner that induced unfolding that was less than this critical level would undergo further unfolding with time and, if so, whether this would induce the onset of platelet adhesion once this critical level was exceeded. To address these questions, CD spectropolarimetry was used to monitor the structure of Alb on OH- and CH(3)-functionalized alkanethiol self-assembled monolayer surfaces, with the Alb initially adsorbed under conditions resulting in degrees of unfolding that were below this critical level, and then the adsorbed Alb layers were aged over 6 months in sterile physiological saline at 37 °C. Platelet adhesion to Alb was quantified at selected time points via a lactate dehydrogenase (LDH) assay. The results indicate that an adsorbed Alb layer does undergo further structural changes with increasing residence time and supports platelet adhesion once it unfolds beyond the previously determined critical level. These results may be relevant to the clinically observed problem of the onset of late-thrombosis, which occurs on cardiovascular implants such as drug-eluting stents.