Merck
  • Immobilization and bioactivity evaluation of FGF-1 and FGF-2 on powdered silicon-doped hydroxyapatite and their scaffolds for bone tissue engineering.
Immobilization and bioactivity evaluation of FGF-1 and FGF-2 on powdered silicon-doped hydroxyapatite and their scaffolds for bone tissue engineering.
Journal of Materials Science. Materials in Medicine
Maria Jose Feito,Rosa Maria Lozano,Maria Alcaide,Cecilia Ramirez-Santillan,Daniel Arcos et al.