Merck
  • Mutagenesis of human acetylcholinesterase. Identification of residues involved in catalytic activity and in polypeptide folding.
Mutagenesis of human acetylcholinesterase. Identification of residues involved in catalytic activity and in polypeptide folding.
The Journal of biological chemistry (1992-09-05)
A Shafferman, C Kronman, Y Flashner, M Leitner, H Grosfeld, A Ordentlich, Y Gozes, S Cohen, N Ariel, D Barak
ABSTRACT
Evidence for the involvement of Ser-203, His-447, and Glu-334 in the catalytic triad of human acetylcholinesterase was provided by substitution of these amino acids by alanine residues. Of 20 amino acid positions mutated so far in human acetylcholinesterase (AChE), these three were unique in abolishing detectable enzymatic activity (less than 0.0003 of wild type), yet allowing proper production, folding, and secretion. This is the first biochemical evidence for the involvement of a glutamate in a hydrolase triad (Schrag, J.D., Li, Y., Wu, M., and Cygler, M. (1991) Nature 351, 761-764), supporting the x-ray crystal structure data of the Torpedo californica acetylcholinesterase (Sussman, J.L., Harel, M., Frolow, F., Oefner, C., Goldman, A., Toker, L. and Silman, I. (1991) Science 253, 872-879). Attempts to convert the AChE triad into a Cys-His-Glu or Ser-His-Asp configuration by site-directed mutagenesis did not yield effective AChE activity. Another type of substitution, that of Asp-74 by Gly or Asn, generated an active enzyme with increased resistance to succinylcholine and dibucaine; thus mimicking in an AChE molecule the phenotype of the atypical butyrylcholinesterase natural variant (D70G mutation). Mutations of other carboxylic residues Glu-84, Asp-95, Asp-333, and Asp-349, all conserved among cholinesterases, did not result in detectable alteration in the recombinant AChE, although polypeptide productivity of the D95N mutant was considerably lower. In contrast, complete absence of secreted human AChE polypeptide was observed when Asp-175 or Asp-404 were substituted by Asn. These two aspartates are conserved in the entire cholinesterase/thyroglobulin family and appear to play a role in generating and/or maintaining the folded state of the polypeptide. The x-ray structure of the Torpedo acetylcholinesterase supports this assumption by revealing the participation of these residues in salt bridges between neighboring secondary structure elements.
Sigma-Aldrich
Acetylcholinesterase human, recombinant, expressed in HEK 293 cells, lyophilized powder, ≥1,000 units/mg protein (BCA)
Sigma-Aldrich
Acetylcholinesterase from human erythrocytes, buffered aqueous solution, ≥500 units/mg protein (BCA)
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type VI-S, lyophilized powder, 200-1,000 units/mg protein