Merck
  • PRC1 and PRC2 are not required for targeting of H2A.Z to developmental genes in embryonic stem cells.
PRC1 and PRC2 are not required for targeting of H2A.Z to developmental genes in embryonic stem cells.
PloS one (2012-04-13)
Robert S Illingworth, Catherine H Botting, Graeme R Grimes, Wendy A Bickmore, Ragnhild Eskeland
ABSTRACT
The essential histone variant H2A.Z localises to both active and silent chromatin sites. In embryonic stem cells (ESCs), H2A.Z is also reported to co-localise with polycomb repressive complex 2 (PRC2) at developmentally silenced genes. The mechanism of H2A.Z targeting is not clear, but a role for the PRC2 component Suz12 has been suggested. Given this association, we wished to determine if polycomb functionally directs H2A.Z incorporation in ESCs. We demonstrate that the PRC1 component Ring1B interacts with multiple complexes in ESCs. Moreover, we show that although the genomic distribution of H2A.Z co-localises with PRC2, Ring1B and with the presence of CpG islands, H2A.Z still blankets polycomb target loci in the absence of Suz12, Eed (PRC2) or Ring1B (PRC1). Therefore we conclude that H2A.Z accumulates at developmentally silenced genes in ESCs in a polycomb independent manner.
Sigma-Aldrich
Gel Filtration Markers Kit for Protein Molecular Weights 29,000-700,000 Da
Sigma-Aldrich
GenomePlex® Complete Whole Genome Amplification (WGA) Kit, Optimized kit with enzyme for amplifying a variety of DNA including FFPE tissue
Sigma-Aldrich
Anti-EZH2 Antibody, Upstate®, from rabbit