Merck
  • The absence of D-alanine from lipoteichoic acid and wall teichoic acid alters surface charge, enhances autolysis and increases susceptibility to methicillin in Bacillus subtilis.
The absence of D-alanine from lipoteichoic acid and wall teichoic acid alters surface charge, enhances autolysis and increases susceptibility to methicillin in <I>Bacillus subtilis</I>.
Microbiology (1997)
Jorg Wecke, Kazimierz Madela, et al.