Merck
  • Genotype-phenotype correlations in pathology caused by collagen type IV alpha 1 and 2 mutations
Genotype-phenotype correlations in pathology caused by collagen type IV alpha 1 and 2 mutations
Matrix Biology (2017)
Jeanne M and Gould DB