Merck
  • Substrate-leash amplification with ribonuclease S-peptide and S-protein.
Substrate-leash amplification with ribonuclease S-peptide and S-protein.
Clinical chemistry (1986-09-01)
M Ehrat, D J Cecchini, R W Giese
ABSTRACT
The S-peptide and S-protein fragments of ribonuclease S (RNase S, no EC no. assigned) have been immobilized onto separate Sepharose gels via a "leash" of polycytidylic acid substrate. Each of these gels releases its RNase fragment when treated with the complementary enzyme fragment or with RNase A (EC 3.1.27.5), and the released fragments recombine to give RNase S activity. Thus this system provides substrate-leash amplification (SLA), such that more enzymatic activity is eluted from the system than is applied. For example, 100 pg of RNase applied to the S-peptide gel is amplified by 1.9 X 10(4) to the equivalent of 1.9 micrograms of activity in 20 h, when followed by combination of the released S-peptide with excess S-protein. We also tested a three-stage amplification system, with a pair of S-peptide and S-protein gels at each stage. In this system the cumulative amplification of the initial 1-ng dose of RNase A is 4.9, 52, and 25-fold after each stage, respectively. Only 2 mg of each SLA gel is used per stage in these experiments, reflecting the magnitude of their production of RNase S activity.